Learn More
Electron Microscopy Sciences Grid Coating Pen For TEM; Coat Quick "G"
SDP
Catalog No. 50930416
Encompass_Preferred
The Coat-Quick "G" pen improves the adherence of tissue sections onto the grids. With a touch of the pen to the grid, a thin layer of coating is applied to the grid. Drying takes place in approximately 1-2 minutes at room temperature. After it has dried the grid is ready for tissue mounting. The pen is also used in pretreating grids prior to mounting supporting films such as formvar and carbon; it minimizes dislodging, widening, or breaking of the support film.
Catalog No. 50-930-416 Supplier Electron Microscopy Sciences Supplier No. 70624
Add to Cart
Add to Cart
The Coat-Quick "G" pen improves the adherence of tissue sections onto the grids. With a touch of the pen to the grid, a thin layer of coating is applied to the grid. Drying takes place in approximately 1-2 minutes at room temperature. After it has dried the grid is ready for tissue mounting. The pen is also used in pretreating grids prior to mounting supporting films such as formvar and carbon; it minimizes dislodging, widening, or breaking of the support film.