Learn More
Electron Microscopy Sciences Grid Dot Array
SDP
Catalog No. 501902454
Encompass_Preferred
The grid dot array is the ideal product for testing image area, distortion, field flatness, and other parameters in optical and imaging systems. Made of green float glass with high reflective chrome image.
The three array areas give options for different magnifications or field size. The R76, a unique product with three dot arrays to suit different magnification or image areas features 3 image areas, with dot size and pitch different in each area. 12 x 9 array of 1 mm dots at 5 mm pitch. 16 x 12 array of 0.5 mm dots at 2 mm pitch. 24 x 18 array of 0.2 mm dots at 1 mm pitch.
Catalog No. 50-190-2454 Supplier Electron Microscopy Sciences Supplier No. 68073R76
The grid dot array is the ideal product for testing image area, distortion, field flatness, and other parameters in optical and imaging systems. Made of green float glass with high reflective chrome image.
The three array areas give options for different magnifications or field size. The R76, a unique product with three dot arrays to suit different magnification or image areas features 3 image areas, with dot size and pitch different in each area. 12 x 9 array of 1 mm dots at 5 mm pitch. 16 x 12 array of 0.5 mm dots at 2 mm pitch. 24 x 18 array of 0.2 mm dots at 1 mm pitch.