Learn More
Electron Microscopy Sciences Gilder Finder Grid F1, 400 Mesh, Copper
SDP
Catalog No. 501927777
Encompass_Preferred
Used to identify specific areas of mesh by an alphabetical or numerical code built into the mesh. 3.05 mm diameter, 0.7 mil (18 um) thickness. The block of 1152 (32x36) individual cells is divided into smaller blocks bounded by thicker crenellated bars. Each small block of 36 (6x6) cells has a central number from 1 to 32 for identification.
Grid: Copper
Catalog No. 50-192-7777 Supplier Electron Microscopy Sciences Supplier No. G400F1CU
Used to identify specific areas of mesh by an alphabetical or numerical code built into the mesh. 3.05 mm diameter, 0.7 mil (18 um) thickness. The block of 1152 (32x36) individual cells is divided into smaller blocks bounded by thicker crenellated bars. Each small block of 36 (6x6) cells has a central number from 1 to 32 for identification.
Grid: Copper