Learn More
Electron Microscopy Sciences Gilder Standard Square 175 mesh Gold Grid
SDP
Catalog No. 501901229
Encompass_Preferred
A reliable support specimen grid source. Features well-defined grid bars, maximum open area, and a matt/shiny side. Each grid is individually inspected. 3.05 mm diameter, 0.7 mil (18 um) thickness.
Grid: Gold
Pitch: 145 µm
Hole: 108 µm
Bar: 37 µm
:
Catalog No. 50-190-1229 Supplier Electron Microscopy Sciences Supplier No. G175AU
A reliable support specimen grid source. Features well-defined grid bars, maximum open area, and a matt/shiny side. Each grid is individually inspected. 3.05 mm diameter, 0.7 mil (18 um) thickness.
Grid: Gold
Pitch: 145 µm
Hole: 108 µm
Bar: 37 µm
:
Product Title