Learn More
Electron Microscopy Sciences Gilder Standard Square 75 mesh Gold Grid
SDP
Catalog No. 501901224
Encompass
A reliable support specimen grid source. Features well-defined grid bars, maximum open area, and a matt/shiny side. Each grid is individually inspected. 3.05 mm diameter, 0.7 mil (18 um) thickness.
Grid: Gold
Pitch: 340 µm
Hole: 280 µm
Bar: 60 µm
:
Catalog No. 50-190-1224 Supplier Electron Microscopy Sciences Supplier No. G75AU
A reliable support specimen grid source. Features well-defined grid bars, maximum open area, and a matt/shiny side. Each grid is individually inspected. 3.05 mm diameter, 0.7 mil (18 um) thickness.
Grid: Gold
Pitch: 340 µm
Hole: 280 µm
Bar: 60 µm
:
Product Title