Learn More
Research Products International Corp Laboratory Notebook, Gridded Pages, Black Cover, 100 Page Book
SDP
Catalog No. 501537761
Encompass
Laboratory notebooks are perfect for recording research data, experimental procedures and design ideas. Use notebooks to help keep information safe and well organized. Hardbound book with imitation leather cover and a title LABORATORY NOTEBOOK foil stampped on the cover. Notebooks feature numbered, acid-free pages that include spaces for title, project number, book number, date and witness signature. A red ribbon marker quickly and easily guides you to where you left off in your notes. Notebook cover is waterproof and chemical-resistant and books contain a periodic and conversion tables in the back.
Catalog No. 50-153-7761 Supplier Research Products International Corp Supplier No. 212612
Laboratory notebooks are perfect for recording research data, experimental procedures and design ideas. Use notebooks to help keep information safe and well organized. Hardbound book with imitation leather cover and a title LABORATORY NOTEBOOK foil stampped on the cover. Notebooks feature numbered, acid-free pages that include spaces for title, project number, book number, date and witness signature. A red ribbon marker quickly and easily guides you to where you left off in your notes. Notebook cover is waterproof and chemical-resistant and books contain a periodic and conversion tables in the back.