Learn More
Electron Microscopy Sciences Gilder Parallel Bar Grid G400P, Gold
SDP
Catalog No. 501901262
Encompass_Preferred
Gilder grids are a reliable support specimen grid for transmission electron microscopy. They feature a well-defined grid bars, maximum open area, and a matt/shiny side. Each grid is individually inspected.
Material: Gold
Pitch: 62 µm
Hole: 22 µm
Bar: 40 µm
Catalog No. 50-190-1262 Supplier Electron Microscopy Sciences Supplier No. G400PAU
Gilder grids are a reliable support specimen grid for transmission electron microscopy. They feature a well-defined grid bars, maximum open area, and a matt/shiny side. Each grid is individually inspected.
Material: Gold
Pitch: 62 µm
Hole: 22 µm
Bar: 40 µm