Learn More
Electron Microscopy Sciences Gilder Parallel Bar Grid G75PB, Gold
SDP
Catalog No. 501901249
Encompass_Preferred
Gilder grids are a reliable support specimen grid for transmission electron microscopy. They feature a well-defined grid bars, maximum open area, and a matt/shiny side. Each grid is individually inspected.
Material: Gold
Pitch: 340 µm
Hole: 270 µm
Bar: 70 µm
Catalog No. 50-190-1249 Supplier Electron Microscopy Sciences Supplier No. G75PBAU
Gilder grids are a reliable support specimen grid for transmission electron microscopy. They feature a well-defined grid bars, maximum open area, and a matt/shiny side. Each grid is individually inspected.
Material: Gold
Pitch: 340 µm
Hole: 270 µm
Bar: 70 µm