Learn More
Electron Microscopy Sciences Gilder Parallel Bar Grid G50P, Nickel
SDP
Catalog No. 501901239
Encompass_Preferred
Gilder grids are a reliable support specimen grid for transmission electron microscopy. They feature a well-defined grid bars, maximum open area, and a matt/shiny side. Each grid is individually inspected.
Material: Nickel
Pitch: 500 µm
Hole: 416 µm
Bar: 84 µm
Catalog No. 50-190-1239 Supplier Electron Microscopy Sciences Supplier No. G50PNI
Gilder grids are a reliable support specimen grid for transmission electron microscopy. They feature a well-defined grid bars, maximum open area, and a matt/shiny side. Each grid is individually inspected.
Material: Nickel
Pitch: 500 µm
Hole: 416 µm
Bar: 84 µm