Learn More
Electron Microscopy Sciences Gilder Parallel Bar Grid G150PB, Copper
SDP
Catalog No. 501901255
Encompass_Preferred
Gilder grids are a reliable support specimen grid for transmission electron microscopy. They feature a well-defined grid bars, maximum open area, and a matt/shiny side. Each grid is individually inspected.
Material: Copper
Pitch: 165 µm
Hole: 115 µm
Bar: 50 µm
Catalog No. 50-190-1255 Supplier Electron Microscopy Sciences Supplier No. G150PBCU
Gilder grids are a reliable support specimen grid for transmission electron microscopy. They feature a well-defined grid bars, maximum open area, and a matt/shiny side. Each grid is individually inspected.
Material: Copper
Pitch: 165 µm
Hole: 115 µm
Bar: 50 µm