Learn More
Electron Microscopy Sciences Gilder Parallel Bar Grid G75PB, Nickel
SDP
Catalog No. 501901248
Encompass_Preferred
Gilder grids are a reliable support specimen grid for transmission electron microscopy. They feature a well-defined grid bars, maximum open area, and a matt/shiny side. Each grid is individually inspected.
Material: Nickel
Pitch: 340 µm
Hole: 270 µm
Bar: 70 µm
Catalog No. 50-190-1248 Supplier Electron Microscopy Sciences Supplier No. G75PBNI
Gilder grids are a reliable support specimen grid for transmission electron microscopy. They feature a well-defined grid bars, maximum open area, and a matt/shiny side. Each grid is individually inspected.
Material: Nickel
Pitch: 340 µm
Hole: 270 µm
Bar: 70 µm