Learn More
Mitegen Grid Prep Block for Cryo-EM Grids, Supports 5 Grids During Surface Treatment
SDP
Catalog No. 5000495031
Encompass_Preferred
The Grid Prep Block is a medical-grade aluminum support block designed to physically support up to five cryo-electron microscopy grids during a variety of surface treatment protocols, with a primary focus on plasma activation. Its ergonomic design includes side finger grooves for a secure grip, numbered support locations for easy tracking, and troughs that provide simple tweezer access both above and below the grid. This product offers a simple, reliable, and efficient means of handling, transporting, and surface-treating grids prior to cryocooling. By providing dedicated support for your grids, the Grid Prep Block helps to streamline your sample preparation workflow and improve the consistency of your results.
Catalog No. 50-004-95031 Supplier Mitegen Supplier No. MCEMGSB51
The Grid Prep Block is a medical-grade aluminum support block designed to physically support up to five cryo-electron microscopy grids during a variety of surface treatment protocols, with a primary focus on plasma activation. Its ergonomic design includes side finger grooves for a secure grip, numbered support locations for easy tracking, and troughs that provide simple tweezer access both above and below the grid. This product offers a simple, reliable, and efficient means of handling, transporting, and surface-treating grids prior to cryocooling. By providing dedicated support for your grids, the Grid Prep Block helps to streamline your sample preparation workflow and improve the consistency of your results.