Learn More
Electron Microscopy Sciences EMS Grid Prep Holder, 81 Capacity
SDP
Catalog No. 501928076
Encompass_Preferred
Unique preparation trays which may be used for staining and/or placing an evaporator for the coating of grids. The trays have a M4 tapped hole in the center which allows for insertion of pin mount. They also have a screw pin in the middle that may be used as a handle for manipulating the trays. They are made from medical grade aluminum and come in a plastic storage box.
Catalog No. 50-192-8076 Supplier Electron Microscopy Sciences Supplier No. 7117502
Unique preparation trays which may be used for staining and/or placing an evaporator for the coating of grids. The trays have a M4 tapped hole in the center which allows for insertion of pin mount. They also have a screw pin in the middle that may be used as a handle for manipulating the trays. They are made from medical grade aluminum and come in a plastic storage box.