Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences Grid Reticle RET 73, 21 mm, 10 mm x 10 mm Scale, 1 mm squares Divisions
SDP

Catalog No. 501901761
Encompass_Preferred

High quality, precision made standard reticles are manufactured to use with our microscope eyepieces. All recticles are chrome on glass and positive polarity.
Size: 21 mm
Scale: 10 mm x 10 mm
Divisions: 1 mm squares
Line width: 25 µm

Catalog No. 50-190-1761 Supplier Electron Microscopy Sciences Supplier No. 6028321
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High quality, precision made standard reticles are manufactured to use with our microscope eyepieces. All recticles are chrome on glass and positive polarity.
Size: 21 mm
Scale: 10 mm x 10 mm
Divisions: 1 mm squares
Line width: 25 µm

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.