Learn More
Fisherbrand™ Grid Reticule
GREENER_CHOICE
Catalog No. 03000132 Shop All Fisherbrand Products
Change view
Click to view available options
For Use With (Equipment):
Microscope
Catalog No. For Use With (Equipment)
03-000-132 Microscope
1 options
Catalog No. 03-000-132
Add to Cart
Add to Cart
Grid reticule with 1 mm squares. Includes spring clip.
  • Reticule grid
  • 10 x 10 mm with 1 mm squares
  • 21 mm diameter with spring clip
Specifications
Type Microscope Eyepiece Reticle
For Use With (Equipment) Microscope