Learn More
Hampton Research GRID SCREEN AMMONIUM SU 185ML
SDP
Catalog No. 501874
Encompass
HR2-924-A3/Grid Screen Ammonium Sulfate #A3/Grid Screen Salt HT (A3), 185 ml - 0.1 M MES monohydrate pH 6.0, 0.8 M Ammonium sulfate. Please note items from this supplier are non-returnable.
Catalog No. 50-187-4 Supplier Hampton Research Supplier No. HR2924A3
HR2-924-A3/Grid Screen Ammonium Sulfate #A3/Grid Screen Salt HT (A3), 185 ml - 0.1 M MES monohydrate pH 6.0, 0.8 M Ammonium sulfate. Please note items from this supplier are non-returnable.