Learn More
Hampton Research HR2-924-B6/Grid Screen Ammonium Sulfate #B6/Grid Screen Salt HT (A12), 185 ml - 0.1 M BICINE pH 9.0, 1.6 M Ammonium sulfate
SDP
Catalog No. 501883
Encompass
HR2-924-B6/Grid Screen Ammonium Sulfate #B6/Grid Screen Salt HT (A12), 185 ml - 0.1 M BICINE pH 9.0, 1.6 M Ammonium sulfate. Please note items from this supplier are non-returnable.
Catalog No. 50-188-3 Supplier Hampton Research Supplier No. HR2924B6
HR2-924-B6/Grid Screen Ammonium Sulfate #B6/Grid Screen Salt HT (A12), 185 ml - 0.1 M BICINE pH 9.0, 1.6 M Ammonium sulfate. Please note items from this supplier are non-returnable.