Learn More
Hampton Research HR2-924-C4/Grid Screen Ammonium Sulfate #C4/Grid Screen Salt HT (B4), 185 ml - 0.1 M HEPES pH 7.0, 2.4 M Ammonium sulfate
SDP
Catalog No. 501887
Encompass
HR2-924-C4/Grid Screen Ammonium Sulfate #C4/Grid Screen Salt HT (B4), 185 ml - 0.1 M HEPES pH 7.0, 2.4 M Ammonium sulfate. Please note items from this supplier are non-returnable.
Catalog No. 50-188-7 Supplier Hampton Research Supplier No. HR2924C4
Add to Cart
Add to Cart
HR2-924-C4/Grid Screen Ammonium Sulfate #C4/Grid Screen Salt HT (B4), 185 ml - 0.1 M HEPES pH 7.0, 2.4 M Ammonium sulfate. Please note items from this supplier are non-returnable.