Learn More
Hampton Research HR2-924-D2/Grid Screen Ammonium Sulfate #D2/Grid Screen Salt HT (B8), 185 ml - 0.1 M Citric acid pH 5.0, 3.0 M Ammonium sulfate
SDP
Catalog No. 501891
Encompass
HR2-924-D2/Grid Screen Ammonium Sulfate #D2/Grid Screen Salt HT (B8), 185 ml - 0.1 M Citric acid pH 5.0, 3.0 M Ammonium sulfate. Please note items from this supplier are non-returnable.
Catalog No. 50-189-1 Supplier Hampton Research Supplier No. HR2924D2
HR2-924-D2/Grid Screen Ammonium Sulfate #D2/Grid Screen Salt HT (B8), 185 ml - 0.1 M Citric acid pH 5.0, 3.0 M Ammonium sulfate. Please note items from this supplier are non-returnable.