Learn More
Hampton Research HR2-924-C6/Grid Screen Ammonium Sulfate #C6/Grid Screen Salt HT (B6), 185 ml - 0.1 M BICINE pH 9.0, 2.4 M Ammonium sulfate
SDP
Catalog No. 501889
Encompass
HR2-924-C6/Grid Screen Ammonium Sulfate #C6/Grid Screen Salt HT (B6), 185 ml - 0.1 M BICINE pH 9.0, 2.4 M Ammonium sulfate. Please note items from this supplier are non-returnable.
Catalog No. 50-188-9 Supplier Hampton Research Supplier No. HR2924C6
HR2-924-C6/Grid Screen Ammonium Sulfate #C6/Grid Screen Salt HT (B6), 185 ml - 0.1 M BICINE pH 9.0, 2.4 M Ammonium sulfate. Please note items from this supplier are non-returnable.