Learn More
Hampton Research GRID SCREEN AMMONIUM SU 185ML
SDP
Catalog No. 501872
Encompass
HR2-924-A1/Grid Screen Ammonium Sulfate #A1/Grid Screen Salt HT (A1), 185 ml - 0.1 M Citric acid pH 4.0, 0.8 M Ammonium sulfate. Please note items from this supplier are non-returnable.
Catalog No. 50-187-2 Supplier Hampton Research Supplier No. HR2924A1
HR2-924-A1/Grid Screen Ammonium Sulfate #A1/Grid Screen Salt HT (A1), 185 ml - 0.1 M Citric acid pH 4.0, 0.8 M Ammonium sulfate. Please note items from this supplier are non-returnable.