Learn More
Hampton Research HR2-924-D3/Grid Screen Ammonium Sulfate #D3/Grid Screen Salt HT (B9), 185 ml - 0.1 M MES monohydrate pH 6.0, 3.0 M Ammonium sulfate
SDP
Catalog No. 501892
Encompass
HR2-924-D3/Grid Screen Ammonium Sulfate #D3/Grid Screen Salt HT (B9), 185 ml - 0.1 M MES monohydrate pH 6.0, 3.0 M Ammonium sulfate. Please note items from this supplier are non-returnable.
Catalog No. 50-189-2 Supplier Hampton Research Supplier No. HR2924D3
Add to Cart
Add to Cart
HR2-924-D3/Grid Screen Ammonium Sulfate #D3/Grid Screen Salt HT (B9), 185 ml - 0.1 M MES monohydrate pH 6.0, 3.0 M Ammonium sulfate. Please note items from this supplier are non-returnable.