Learn More
Hampton Research HR2-924-D1/Grid Screen Ammonium Sulfate #D1/Grid Screen Salt HT (B7), 185 ml - 0.1 M Citric acid pH 4.0, 3.0 M Ammonium sulfate
SDP
Catalog No. 501890
Encompass
HR2-924-D1/Grid Screen Ammonium Sulfate #D1/Grid Screen Salt HT (B7), 185 ml - 0.1 M Citric acid pH 4.0, 3.0 M Ammonium sulfate. Please note items from this supplier are non-returnable.
Catalog No. 50-189-0 Supplier Hampton Research Supplier No. HR2924D1
Add to Cart
Add to Cart
HR2-924-D1/Grid Screen Ammonium Sulfate #D1/Grid Screen Salt HT (B7), 185 ml - 0.1 M Citric acid pH 4.0, 3.0 M Ammonium sulfate. Please note items from this supplier are non-returnable.