Learn More
Hampton Research HR2-924-D4/Grid Screen Ammonium Sulfate #D4/Grid Screen Salt HT (B10), 185 ml - 0.1 M HEPES pH 7.0, 3.0 M Ammonium sulfate
SDP
Catalog No. 501893
Encompass
HR2-924-D4/Grid Screen Ammonium Sulfate #D4/Grid Screen Salt HT (B10), 185 ml - 0.1 M HEPES pH 7.0, 3.0 M Ammonium sulfate. Please note items from this supplier are non-returnable.
Catalog No. 50-189-3 Supplier Hampton Research Supplier No. HR2924D4
HR2-924-D4/Grid Screen Ammonium Sulfate #D4/Grid Screen Salt HT (B10), 185 ml - 0.1 M HEPES pH 7.0, 3.0 M Ammonium sulfate. Please note items from this supplier are non-returnable.