Learn More
Hampton Research GRID SCREEN AMMONIUM SU 185ML
SDP
Catalog No. 501873
Encompass
HR2-924-A2/Grid Screen Ammonium Sulfate #A2/Grid Screen Salt HT (A2), 185 ml - 0.1 M Citric acid pH 5.0, 0.8 M Ammonium sulfate. Please note items from this supplier are non-returnable.
Catalog No. 50-187-3 Supplier Hampton Research Supplier No. HR2924A2
HR2-924-A2/Grid Screen Ammonium Sulfate #A2/Grid Screen Salt HT (A2), 185 ml - 0.1 M Citric acid pH 5.0, 0.8 M Ammonium sulfate. Please note items from this supplier are non-returnable.