Learn More
Hampton Research HR2-924-B5/Grid Screen Ammonium Sulfate #B5/Grid Screen Salt HT (A11), 185 ml - 0.1 M Tris pH 8.0, 1.6 M Ammonium sulfate
SDP
Catalog No. 501882
Encompass
HR2-924-B5/Grid Screen Ammonium Sulfate #B5/Grid Screen Salt HT (A11), 185 ml - 0.1 M Tris pH 8.0, 1.6 M Ammonium sulfate. Please note items from this supplier are non-returnable.
Catalog No. 50-188-2 Supplier Hampton Research Supplier No. HR2924B5
HR2-924-B5/Grid Screen Ammonium Sulfate #B5/Grid Screen Salt HT (A11), 185 ml - 0.1 M Tris pH 8.0, 1.6 M Ammonium sulfate. Please note items from this supplier are non-returnable.