Learn More
Hampton Research HR2-924-C5/Grid Screen Ammonium Sulfate #C5/Grid Screen Salt HT (B5), 185 ml - 0.1 M Tris pH 8.0, 2.4 M Ammonium sulfate
SDP
Catalog No. 501888
Encompass
HR2-924-C5/Grid Screen Ammonium Sulfate #C5/Grid Screen Salt HT (B5), 185 ml - 0.1 M Tris pH 8.0, 2.4 M Ammonium sulfate. Please note items from this supplier are non-returnable.
Catalog No. 50-188-8 Supplier Hampton Research Supplier No. HR2924C5
HR2-924-C5/Grid Screen Ammonium Sulfate #C5/Grid Screen Salt HT (B5), 185 ml - 0.1 M Tris pH 8.0, 2.4 M Ammonium sulfate. Please note items from this supplier are non-returnable.