Learn More
Hampton Research HR2-924-A4/Grid Screen Ammonium Sulfate #A4/Grid Screen Salt HT (A4), 185 ml - 0.1 M HEPES pH 7.0, 0.8 M Ammonium sulfate
SDP
Catalog No. 501875
Encompass
HR2-924-A4/Grid Screen Ammonium Sulfate #A4/Grid Screen Salt HT (A4), 185 ml - 0.1 M HEPES pH 7.0, 0.8 M Ammonium sulfate. Please note items from this supplier are non-returnable.
Catalog No. 50-187-5 Supplier Hampton Research Supplier No. HR2924A4
Add to Cart
Add to Cart
HR2-924-A4/Grid Screen Ammonium Sulfate #A4/Grid Screen Salt HT (A4), 185 ml - 0.1 M HEPES pH 7.0, 0.8 M Ammonium sulfate. Please note items from this supplier are non-returnable.