Learn More
Hampton Research HR2-924-B4/Grid Screen Ammonium Sulfate #B4/Grid Screen Salt HT (A10), 185 ml - 0.1 M HEPES pH 7.0, 1.6 M Ammonium sulfate
SDP
Supplier:  Hampton Research HR2924B4
Encompass
HR2-924-B4/Grid Screen Ammonium Sulfate #B4/Grid Screen Salt HT (A10), 185 ml - 0.1 M HEPES pH 7.0, 1.6 M Ammonium sulfate. Please note items from this supplier are non-returnable.
Catalog No. 50-188-1