Learn More
Hampton Research HR2-930-A2/Grid Screen MPD #A2, 185 ml - 0.1 M Sodium acetate trihydrate pH 5.0, 10% (+/-)-2-Methyl-2,4-pentanediol
SDP
Catalog No. 501945
Encompass
HR2-930-A2/Grid Screen MPD #A2, 185 ml - 0.1 M Sodium acetate trihydrate pH 5.0, 10% (+/-)-2-Methyl-2,4-pentanediol. Please note items from this supplier are non-returnable.
Catalog No. 50-194-5 Supplier Hampton Research Supplier No. HR2930A2
HR2-930-A2/Grid Screen MPD #A2, 185 ml - 0.1 M Sodium acetate trihydrate pH 5.0, 10% (+/-)-2-Methyl-2,4-pentanediol. Please note items from this supplier are non-returnable.