Learn More
Hampton Research HR2-930-B2/Grid Screen MPD #B2, 185 ml - 0.1 M Sodium acetate trihydrate pH 5.0, 20% (+/-)-2-Methyl-2,4-pentanediol
SDP
Catalog No. 501951
Encompass
HR2-930-B2/Grid Screen MPD #B2, 185 ml - 0.1 M Sodium acetate trihydrate pH 5.0, 20% (+/-)-2-Methyl-2,4-pentanediol. Please note items from this supplier are non-returnable.
Catalog No. 50-195-1 Supplier Hampton Research Supplier No. HR2930B2
HR2-930-B2/Grid Screen MPD #B2, 185 ml - 0.1 M Sodium acetate trihydrate pH 5.0, 20% (+/-)-2-Methyl-2,4-pentanediol. Please note items from this supplier are non-returnable.