Learn More
Hampton Research HR2-930-C3/Grid Screen MPD #C3, 185 ml - 0.1 M MES monohydrate pH 6.0, 40% (+/-)-2-Methyl-2,4-pentanediol
SDP
Catalog No. 501958
Encompass
HR2-930-C3/Grid Screen MPD #C3, 185 ml - 0.1 M MES monohydrate pH 6.0, 40% (+/-)-2-Methyl-2,4-pentanediol. Please note items from this supplier are non-returnable.
Catalog No. 50-195-8 Supplier Hampton Research Supplier No. HR2930C3
HR2-930-C3/Grid Screen MPD #C3, 185 ml - 0.1 M MES monohydrate pH 6.0, 40% (+/-)-2-Methyl-2,4-pentanediol. Please note items from this supplier are non-returnable.