Learn More
Hampton Research HR2-930-C4/Grid Screen MPD #C4, 185 ml - 0.1 M HEPES pH 7.0, 40% (+/-)-2-Methyl-2,4-pentanediol
SDP
Catalog No. 501959
Encompass
HR2-930-C4/Grid Screen MPD #C4, 185 ml - 0.1 M HEPES pH 7.0, 40% (+/-)-2-Methyl-2,4-pentanediol. Please note items from this supplier are non-returnable.
Catalog No. 50-195-9 Supplier Hampton Research Supplier No. HR2930C4
HR2-930-C4/Grid Screen MPD #C4, 185 ml - 0.1 M HEPES pH 7.0, 40% (+/-)-2-Methyl-2,4-pentanediol. Please note items from this supplier are non-returnable.