Learn More
Hampton Research HR2-926-A2/Grid Screen PEG 6000 #A2, 185 ml - 0.1 M Citric acid pH 5.0, 5% w/v Polyethylene glycol 6,000
SDP
Catalog No. 501897
Encompass
HR2-926-A2/Grid Screen PEG 6000 #A2, 185 ml - 0.1 M Citric acid pH 5.0, 5% w/v Polyethylene glycol 6,000. Please note items from this supplier are non-returnable.
Catalog No. 50-189-7 Supplier Hampton Research Supplier No. HR2926A2
HR2-926-A2/Grid Screen PEG 6000 #A2, 185 ml - 0.1 M Citric acid pH 5.0, 5% w/v Polyethylene glycol 6,000. Please note items from this supplier are non-returnable.