Learn More
Hampton Research HR2-926-B4/Grid Screen PEG 6000 #B4, 185 ml - 0.1 M HEPES pH 7.0, 10% w/v Polyethylene glycol 6,000
SDP
Catalog No. 501905
Encompass
HR2-926-B4/Grid Screen PEG 6000 #B4, 185 ml - 0.1 M HEPES pH 7.0, 10% w/v Polyethylene glycol 6,000. Please note items from this supplier are non-returnable.
Catalog No. 50-190-5 Supplier Hampton Research Supplier No. HR2926B4
HR2-926-B4/Grid Screen PEG 6000 #B4, 185 ml - 0.1 M HEPES pH 7.0, 10% w/v Polyethylene glycol 6,000. Please note items from this supplier are non-returnable.