Learn More
Hampton Research HR2-926-B6/Grid Screen PEG 6000 #B6, 185 ml - 0.1 M BICINE pH 9.0, 10% w/v Polyethylene glycol 6,000
SDP
Catalog No. 501907
Encompass
HR2-926-B6/Grid Screen PEG 6000 #B6, 185 ml - 0.1 M BICINE pH 9.0, 10% w/v Polyethylene glycol 6,000. Please note items from this supplier are non-returnable.
Catalog No. 50-190-7 Supplier Hampton Research Supplier No. HR2926B6
HR2-926-B6/Grid Screen PEG 6000 #B6, 185 ml - 0.1 M BICINE pH 9.0, 10% w/v Polyethylene glycol 6,000. Please note items from this supplier are non-returnable.