Learn More
Hampton Research HR2-926-C2/Grid Screen PEG 6000 #C2, 185 ml - 0.1 M Citric acid pH 5.0, 20% w/v Polyethylene glycol 6,000
SDP
Catalog No. 501909
Encompass
HR2-926-C2/Grid Screen PEG 6000 #C2, 185 ml - 0.1 M Citric acid pH 5.0, 20% w/v Polyethylene glycol 6,000. Please note items from this supplier are non-returnable.
Catalog No. 50-190-9 Supplier Hampton Research Supplier No. HR2926C2
HR2-926-C2/Grid Screen PEG 6000 #C2, 185 ml - 0.1 M Citric acid pH 5.0, 20% w/v Polyethylene glycol 6,000. Please note items from this supplier are non-returnable.