Learn More
Hampton Research HR2-926-C4/Grid Screen PEG 6000 #C4, 185 ml - 0.1 M HEPES pH 7.0, 20% w/v Polyethylene glycol 6,000
SDP
Catalog No. 501911
Encompass
HR2-926-C4/Grid Screen PEG 6000 #C4, 185 ml - 0.1 M HEPES pH 7.0, 20% w/v Polyethylene glycol 6,000. Please note items from this supplier are non-returnable.
Catalog No. 50-191-1 Supplier Hampton Research Supplier No. HR2926C4
HR2-926-C4/Grid Screen PEG 6000 #C4, 185 ml - 0.1 M HEPES pH 7.0, 20% w/v Polyethylene glycol 6,000. Please note items from this supplier are non-returnable.