Learn More
Hampton Research HR2-926-C1/Grid Screen PEG 6000 #C1, 185 ml - 0.1 M Citric acid pH 4.0, 20% w/v Polyethylene glycol 6,000
SDP
Catalog No. 501908
Encompass
HR2-926-C1/Grid Screen PEG 6000 #C1, 185 ml - 0.1 M Citric acid pH 4.0, 20% w/v Polyethylene glycol 6,000. Please note items from this supplier are non-returnable.
Catalog No. 50-190-8 Supplier Hampton Research Supplier No. HR2926C1
HR2-926-C1/Grid Screen PEG 6000 #C1, 185 ml - 0.1 M Citric acid pH 4.0, 20% w/v Polyethylene glycol 6,000. Please note items from this supplier are non-returnable.