Learn More
Hampton Research HR2-926-C6/Grid Screen PEG 6000 #C6, 185 ml - 0.1 M BICINE pH 9.0, 20% w/v Polyethylene glycol 6,000
SDP
Catalog No. 501913
Encompass
HR2-926-C6/Grid Screen PEG 6000 #C6, 185 ml - 0.1 M BICINE pH 9.0, 20% w/v Polyethylene glycol 6,000. Please note items from this supplier are non-returnable.
Catalog No. 50-191-3 Supplier Hampton Research Supplier No. HR2926C6
HR2-926-C6/Grid Screen PEG 6000 #C6, 185 ml - 0.1 M BICINE pH 9.0, 20% w/v Polyethylene glycol 6,000. Please note items from this supplier are non-returnable.