Learn More
Hampton Research HR2-926-D3/Grid Screen PEG 6000 #D3, 185 ml - 0.1 M MES monohydrate pH 6.0, 30% w/v Polyethylene glycol 6,000
SDP
Catalog No. 501916
Encompass
HR2-926-D3/Grid Screen PEG 6000 #D3, 185 ml - 0.1 M MES monohydrate pH 6.0, 30% w/v Polyethylene glycol 6,000. Please note items from this supplier are non-returnable.
Catalog No. 50-191-6 Supplier Hampton Research Supplier No. HR2926D3
HR2-926-D3/Grid Screen PEG 6000 #D3, 185 ml - 0.1 M MES monohydrate pH 6.0, 30% w/v Polyethylene glycol 6,000. Please note items from this supplier are non-returnable.