Learn More
Hampton Research HR2-926-D5/Grid Screen PEG 6000 #D5, 185 ml - 0.1 M Tris pH 8.0, 30% w/v Polyethylene glycol 6,000
SDP
Catalog No. 501918
Encompass
HR2-926-D5/Grid Screen PEG 6000 #D5, 185 ml - 0.1 M Tris pH 8.0, 30% w/v Polyethylene glycol 6,000. Please note items from this supplier are non-returnable.
Catalog No. 50-191-8 Supplier Hampton Research Supplier No. HR2926D5
HR2-926-D5/Grid Screen PEG 6000 #D5, 185 ml - 0.1 M Tris pH 8.0, 30% w/v Polyethylene glycol 6,000. Please note items from this supplier are non-returnable.