Learn More
Hampton Research HR2-928-A3/Grid Screen PEG/LiCl #A3, 185 ml - 0.1 M MES monohydrate pH 6.0, 1.0 M Lithium chloride
SDP
Catalog No. 501922
Encompass
HR2-928-A3/Grid Screen PEG/LiCl #A3, 185 ml - 0.1 M MES monohydrate pH 6.0, 1.0 M Lithium chloride. Please note items from this supplier are non-returnable.
Catalog No. 50-192-2 Supplier Hampton Research Supplier No. HR2928A3
HR2-928-A3/Grid Screen PEG/LiCl #A3, 185 ml - 0.1 M MES monohydrate pH 6.0, 1.0 M Lithium chloride. Please note items from this supplier are non-returnable.