Learn More
Hampton Research HR2-928-A2/Grid Screen PEG/LiCl #A2, 185 ml - 0.1 M Citric acid pH 5.0, 1.0 M Lithium chloride
SDP
Catalog No. 501921
Encompass
HR2-928-A2/Grid Screen PEG/LiCl #A2, 185 ml - 0.1 M Citric acid pH 5.0, 1.0 M Lithium chloride. Please note items from this supplier are non-returnable.
Catalog No. 50-192-1 Supplier Hampton Research Supplier No. HR2928A2
HR2-928-A2/Grid Screen PEG/LiCl #A2, 185 ml - 0.1 M Citric acid pH 5.0, 1.0 M Lithium chloride. Please note items from this supplier are non-returnable.