Learn More
Hampton Research HR2-928-B4/Grid Screen PEG/LiCl #B4, 185 ml - 0.1 M HEPES pH 7.0, 1.0 M Lithium chloride, 10% w/v Polyethylene glycol 6,000
SDP
Catalog No. 501929
Encompass
HR2-928-B4/Grid Screen PEG/LiCl #B4, 185 ml - 0.1 M HEPES pH 7.0, 1.0 M Lithium chloride, 10% w/v Polyethylene glycol 6,000. Please note items from this supplier are non-returnable.
Catalog No. 50-192-9 Supplier Hampton Research Supplier No. HR2928B4
HR2-928-B4/Grid Screen PEG/LiCl #B4, 185 ml - 0.1 M HEPES pH 7.0, 1.0 M Lithium chloride, 10% w/v Polyethylene glycol 6,000. Please note items from this supplier are non-returnable.