Learn More
Hampton Research HR2-928-B1/Grid Screen PEG/LiCl #B1, 185 ml - 0.1 M Citric acid pH 4.0, 1.0 M Lithium chloride, 10% w/v Polyethylene glycol 6,000
SDP
Catalog No. 501926
Encompass
HR2-928-B1/Grid Screen PEG/LiCl #B1, 185 ml - 0.1 M Citric acid pH 4.0, 1.0 M Lithium chloride, 10% w/v Polyethylene glycol 6,000. Please note items from this supplier are non-returnable.
Catalog No. 50-192-6 Supplier Hampton Research Supplier No. HR2928B1
HR2-928-B1/Grid Screen PEG/LiCl #B1, 185 ml - 0.1 M Citric acid pH 4.0, 1.0 M Lithium chloride, 10% w/v Polyethylene glycol 6,000. Please note items from this supplier are non-returnable.