Learn More
Hampton Research HR2-928-B3/Grid Screen PEG/LiCl #B3, 185 ml - 0.1 M MES monohydrate pH 6.0, 1.0 M Lithium chloride, 10% w/v Polyethylene glycol 6,000
SDP
Catalog No. 501928
Encompass
HR2-928-B3/Grid Screen PEG/LiCl #B3, 185 ml - 0.1 M MES monohydrate pH 6.0, 1.0 M Lithium chloride, 10% w/v Polyethylene glycol 6,000. Please note items from this supplier are non-returnable.
Catalog No. 50-192-8 Supplier Hampton Research Supplier No. HR2928B3
HR2-928-B3/Grid Screen PEG/LiCl #B3, 185 ml - 0.1 M MES monohydrate pH 6.0, 1.0 M Lithium chloride, 10% w/v Polyethylene glycol 6,000. Please note items from this supplier are non-returnable.