Learn More
Hampton Research HR2-928-C6/Grid Screen PEG/LiCl #C6, 185 ml - 0.1 M BICINE pH 9.0, 1.0 M Lithium chloride, 20% w/v Polyethylene glycol 6,000
SDP
Catalog No. 501937
Encompass
HR2-928-C6/Grid Screen PEG/LiCl #C6, 185 ml - 0.1 M BICINE pH 9.0, 1.0 M Lithium chloride, 20% w/v Polyethylene glycol 6,000. Please note items from this supplier are non-returnable.
Catalog No. 50-193-7 Supplier Hampton Research Supplier No. HR2928C6
HR2-928-C6/Grid Screen PEG/LiCl #C6, 185 ml - 0.1 M BICINE pH 9.0, 1.0 M Lithium chloride, 20% w/v Polyethylene glycol 6,000. Please note items from this supplier are non-returnable.