Learn More
Hampton Research HR2-928-C3/Grid Screen PEG/LiCl #C3, 185 ml - 0.1 M MES monohydrate pH 6.0, 1.0 M Lithium chloride, 20% w/v Polyethylene glycol 6,000
SDP
Catalog No. 501934
Encompass
HR2-928-C3/Grid Screen PEG/LiCl #C3, 185 ml - 0.1 M MES monohydrate pH 6.0, 1.0 M Lithium chloride, 20% w/v Polyethylene glycol 6,000. Please note items from this supplier are non-returnable.
Catalog No. 50-193-4 Supplier Hampton Research Supplier No. HR2928C3
HR2-928-C3/Grid Screen PEG/LiCl #C3, 185 ml - 0.1 M MES monohydrate pH 6.0, 1.0 M Lithium chloride, 20% w/v Polyethylene glycol 6,000. Please note items from this supplier are non-returnable.