Learn More
Hampton Research HR2-928-D2/Grid Screen PEG/LiCl #D2, 185 ml - 0.1 M Citric acid pH 5.0, 1.0 M Lithium chloride, 30% w/v Polyethylene glycol 6,000
SDP
Catalog No. 501939
Encompass
HR2-928-D2/Grid Screen PEG/LiCl #D2, 185 ml - 0.1 M Citric acid pH 5.0, 1.0 M Lithium chloride, 30% w/v Polyethylene glycol 6,000. Please note items from this supplier are non-returnable.
Catalog No. 50-193-9 Supplier Hampton Research Supplier No. HR2928D2
HR2-928-D2/Grid Screen PEG/LiCl #D2, 185 ml - 0.1 M Citric acid pH 5.0, 1.0 M Lithium chloride, 30% w/v Polyethylene glycol 6,000. Please note items from this supplier are non-returnable.